EMAIL THIS PAGE TO A FRIEND
EMBO molecular medicine
Src-dependent phosphorylation of μ-opioid receptor at Tyr
PMID 28818835
Abstract
Opiate withdrawal/negative reinforcement has been implicated as one of the mechanisms for the progression from impulsive to compulsive drug use. Increase in the intracellular cAMP level and protein kinase A (PKA) activities within the neurocircuitry of addiction has been a leading hypothesis for opiate addiction. This increase requires the phosphorylation of μ-opioid receptor (MOR) at Tyr