EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Mediated water electrolysis in biphasic systems.
PMID 28820214
Abstract
The concept of efficient electrolysis by linking photoelectrochemical biphasic H