EMAIL THIS PAGE TO A FRIEND
Biochemistry
Pathogenic Mutations Induce Partial Structural Changes in the Native β-Sheet Structure of Transthyretin and Accelerate Aggregation.
PMID 28820582
Abstract
Amyloid formation of natively folded proteins involves global and/or local unfolding of the native state to form aggregation-prone intermediates. Here we report solid-state nuclear magnetic resonance (NMR) structural studies of amyloid derived from wild-type (WT) and more aggressive mutant forms of transthyretin (TTR) to investigate the structural changes associated with effective TTR aggregation. We employed selective