EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Comparative study of single/combination use of Huang-Lian-Jie-Du decoction and berberine on their protection on sepsis induced acute liver injury by NMR metabolic profiling.
PMID 28822346
Abstract
Sepsis is a serious clinical disease with a high mortality rate all around the world. Liver organ dysfunction is an important sign for the severity and outcome of sepsis in patients. In this study,