EMAIL THIS PAGE TO A FRIEND
Acta biomaterialia
A pH-sensitive methenamine mandelate-loaded nanoparticle induces DNA damage and apoptosis of cancer cells.
PMID 28822844
Abstract
Methenamine mandelate is a urinary antibacterial agent, which can be converted to formaldehyde in urine that has a relatively low pH of average 5.5-6.8. Here, we prepare a pH-sensitive PLGA-based nanoparticle containing both methenamine mandelate and NaHCO Methenamine mandelate is a urinary antibacterial agent, which can be converted to formaldehyde in urine that has a relatively low pH of average 5.5-6.8. Here, we prepare a pH-sensitive PLGA-based nanoparticle containing both methenamine mandelate and NaHCO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4292
3,3′-Dioctadecyloxacarbocyanine perchlorate
C53H85ClN2O6