EMAIL THIS PAGE TO A FRIEND
Journal of nanoparticle research : an interdisciplinary forum for nanoscale science and technology
Effects of surface functionalization of hydrophilic NaYF
PMID 28824289
Abstract
Specific rare earth doped nanocrystals (NCs), a recent class of nanoparticles with fluorescent features, have great bioanalytical potential. Neuroactive properties of NaYF