EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
Capsule Enlargement in
PMID 28824559
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
292648
Dihydrofluorescein diacetate, 97%
C24H18O7
S607
Salicylhydroxamic acid, 99%
C7H7NO3