EMAIL THIS PAGE TO A FRIEND
Frontiers in plant science
The
PMID 28824679
Abstract
Salinity threatens productivity of economically important crops such as tomato (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z0164
Zeatin, BioReagent, plant cell culture tested, powder
C10H13N5O