EMAIL THIS PAGE TO A FRIEND
Theranostics
Al
PMID 28824726
Abstract
Positron emission tomography (PET) using radiolabeled biomolecules is a translational molecular imaging technology that is increasingly used in support of drug development. Current methods for radiolabeling biomolecules with fluorine-18 are laborious and require multistep procedures with moderate labeling yields. The Al