EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry letters
Li-O
PMID 28825835
Abstract
Redox mediators (RMs) have been widely applied to reduce the charge overpotential of nonaqueous lithium-oxygen (Li-O
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
110175
Tetraethylene glycol, 99%
C8H18O5