EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Advanced glycation end products receptor RAGE controls myocardial dysfunction and oxidative stress in high-fat fed mice by sustaining mitochondrial dynamics and autophagy-lysosome pathway.
PMID 28826719
Abstract
Oxidative stress and mitochondrial dysfunction are recognized as major contributors of cardiovascular damage in diabetes and high fat diet (HFD) fed mice. Blockade of receptor for advanced glycation end products (RAGE) attenuates vascular oxidative stress and development of atherosclerosis. We tested whether HFD-induced myocardial dysfunction would be reversed in RAGE deficiency mice, in association with changes in oxidative stress damage, mitochondrial respiration, mitochondrial fission and autophagy-lysosomal pathway. Cardiac antioxidant capacity was upregulated in RAGE