EMAIL THIS PAGE TO A FRIEND
Plant physiology
TransDetect Identifies a New Regulatory Module Controlling Phosphate Accumulation.
PMID 28827455
Abstract
Identifying transcription factor (TFs) cooperation controlling target gene expression is still an arduous challenge. The accuracy of current methods at genome scale significantly drops with the increase in number of genes, which limits their applicability to more complex genomes, like animals and plants. Here, we developed an algorithm, TransDetect, able to predict TF combinations controlling the expression level of a given gene. TransDetect was used to identify novel TF modules regulating the expression of Arabidopsis (
Related Materials