EMAIL THIS PAGE TO A FRIEND
Scientific reports
PDGF-BB enhances collagen gel contraction through a PI3K-PLCγ-PKC-cofilin pathway.
PMID 28827622
Abstract
Cell-mediated contraction of collagenous matrices is modulated by various growth factors and cytokines, such as platelet-derived growth factor-BB (PDGF-BB). Here we used a genetic cell model to delineate defined signaling pathways that enhance collagen gel contraction downstream of ligand-stimulated platelet-derived growth factor receptor-β (PDGF-Rβ). Our data show that PDGF BB-enhanced activations of phosphatidylinositol 3'-kinase (PI3K) and phospholipase Cγ (PLCγ) were necessary for PDGF-enhanced collagen gel contraction. Importantly, other defined signaling pathways down-stream of PDGF-Rβ were, however, dispensable. The decisive roles for PI3K and PLCγ were corroborated by experiments using selective inhibitors. Furthermore, we show that de-phosphorylation and thereby activation of cofilin that is important for the turnover of actin filaments, is depended on PI3K and PLCγ down-stream of PDGF-Rβ. Moreover, inhibition of protein kinase C (PKC) by GÖ6976 and bisindolylmaleimide-II abolished cofilin de-phosphorylation, as well as PDGF-enhanced contraction. In contrast, activation of the PKC protein family by 4β-phorbol 12-myristate 13-acetate (PMA) did not accelerate collagen gel contraction although it induced long-term cofilin de-phosphorylation, showing the need of a dynamic control of cofilin de-phosphorylation for PDGF-enhanced collagen gel contraction. Taken together, our data point to the involvement of a PI3K/PLCγ-PKC-cofilin pathway in both PDGF-enhanced cofilin de-phosphorylation and PDGF-enhanced collagen gel contraction.