EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Mitotane (1-(o-chlorophenyl)-1-(p-chlorophenyl)-2,2-dichloroethane) metabolism in perfusion studies with dog adrenal glands.
PMID 2882989
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35484
4,4′-DDA, PESTANAL®, analytical standard
C14H10Cl2O2
100870
Bis(4-chlorophenyl)acetic acid, 98%
C14H10Cl2O2