EMAIL THIS PAGE TO A FRIEND
The American journal of Chinese medicine
Syk-Mediated Suppression of Inflammatory Responses by Cordyceps bassiana.
PMID 28830213
Abstract
The fruit body of artificially cultivated Cordyceps bassiana has been reported to exhibit anti-inflammatory and anticancer activities. Although it has been suggested that the fruit body has neutraceutic and pharmaceutic biomaterial potential, the exact anti-inflammatory molecular mechanism has not been fully elucidated. In this study, we demonstrated the immunopharmacologic activity of Cordyceps bassiana under in vitro conditions and investigated its anti-inflammatory mechanism. Water extract (Cm-WE) of the fruit body of artificially cultivated Cordyceps bassiana without polysaccharide fractions reduced the expression of the proinflammatory genes cyclooxygenase (COX)-2, interleukin (IL)-12, and inducible nitric oxide synthase (iNOS) and promoted the expression of the anti-inflammatory gene IL-10 in lipopolysaccharide (LPS)-treated RAW264.7 cells. In addition, this fraction suppressed proliferation and interferon (IFN)-[Formula: see text] production in splenic T lymphocytes. Cm-WE blocked the activation of nuclear factor (NF)-[Formula: see text]B and activator protein (AP)-1 and their upstream inflammatory signaling cascades, including Syk, MEK, and JNK. Using kinase assays, Syk was identified as the target enzyme most strongly inhibited by Cm-WE. These results strongly suggest that Cm-WE suppresses inflammatory responses by inhibiting Syk kinase activity, with potential implications for novel neutraceutic and pharmaceutic biomaterials.