EMAIL THIS PAGE TO A FRIEND
Journal of nanobiotechnology
Characteristics and properties of nano-LiCoO
PMID 28830490
Abstract
LiCoO Several new single source precursors containing lithium (Li Our heterobimetallic single source precursors allow to dramatically reduce the production temperature and time for HT-LCO. The obtained nanoparticles of LiCoO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
400378
Lithium phenoxide solution, 1.0 M in THF
C6H5LiO