EMAIL THIS PAGE TO A FRIEND
Mechanisms of ageing and development
Senescent intervertebral disc cells exhibit perturbed matrix homeostasis phenotype.
PMID 28830687
Abstract
Aging greatly increases the risk for intervertebral disc degeneration (IDD) as a result of proteoglycan loss due to reduced synthesis and enhanced degradation of the disc matrix proteoglycan (PG). How disc matrix PG homeostasis becomes perturbed with age is not known. The goal of this study is to determine whether cellular senescence is a source of this perturbation. We demonstrated that disc cellular senescence is dramatically increased in the DNA repair-deficient Ercc1
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T9039 Tris Buffered Saline, with Tween® 20, pH 8.0, powder