EMAIL THIS PAGE TO A FRIEND
Annals of neurology
Inhibiting persistent inward sodium currents prevents myotonia.
PMID 28833464
Abstract
Patients with myotonia congenita have muscle hyperexcitability due to loss-of-function mutations in the ClC-1 chloride channel in skeletal muscle, which causes involuntary firing of muscle action potentials (myotonia), producing muscle stiffness. The excitatory events that trigger myotonic action potentials in the absence of stabilizing ClC-1 current are not fully understood. Our goal was to identify currents that trigger spontaneous firing of muscle in the setting of reduced ClC-1 current. In vitro intracellular current clamp and voltage clamp recordings were performed in muscle from a mouse model of myotonia congenita. Intracellular recordings revealed a slow afterdepolarization (AfD) that triggers myotonic action potentials. The AfD is well explained by a tetrodotoxin-sensitive and voltage-dependent Na This work significantly changes our understanding of the mechanisms triggering myotonia. Our work suggests that the current focus of treating myotonia, blocking the transient Na