EMAIL THIS PAGE TO A FRIEND
EMBO molecular medicine
Genome editing for scalable production of alloantigen-free lentiviral vectors for
PMID 28835507
Abstract
Lentiviral vectors (LV) are powerful and versatile vehicles for gene therapy. However, their complex biological composition challenges large-scale manufacturing and raises concerns for