EMAIL THIS PAGE TO A FRIEND
Pharmacognosy magazine
Anticancer Potential of Steviol in MCF-7 Human Breast Cancer Cells.
PMID 28839355
Abstract
This study aimed to investigate the cytotoxicity, apoptosis induction, and mechanism of action of steviol on human breast cancer cells (Michigan Cancer Foundation-7 [MCF-7]). Sulforhodamine-B assay was performed to analyze cytotoxic potential of Steviol whereas flow cytometer was used to analyze cell cycle, apoptosis, and reactive oxygen species generation. Studying the viability of cells confirms the IC Thus, results suggest that induction of apoptosis in MCF-7 cells was due to dose-dependent effect of Steviol. Our first Steviol remarkably inhibited the growth MCF-7 HBCCs in a dose dependent mannerIt abolishes cell cycle progression by arresting cells at G2/M phaseSteviol induces the cells to undergo apoptosisSteviol induces the cells to generate reactive oxygen species (ROS).