EMAIL THIS PAGE TO A FRIEND
Talanta
A novel high throughput screening assay for binding affinities of perfluoroalkyl iodide for estrogen receptor alpha and beta isoforms.
PMID 28842010
Abstract
Contaminants of emerging concern are continuously increasing, which makes it important to develop high throughput screening techniques for the evaluation of their potential biological effects, especially endocrine disrupting effects, which would directly influence the population dynamics in environment. A novel competitive binding assay based on enzyme fragmentation complementation technology was established to screen the binding affinities of emerging chemicals for estrogen receptor (ER) α or β isoforms. Exogenous compounds could compete with the fragment (ED-ES) of genetically engineered β-galactosidase enzyme (β-gal) for the binding to ERα or β, thus quantitatively altering the formation of enzymatically active β-gal and the hydrolysis of luminescent substrate. According to the monitoring of luminescence curves and the optimization of ERα or β concentrations, it was found that luminescent signals were sustainably emitted for 9h, and 40nM ERα or β in the system would lead to the most sensitive luminescence response. Using 17β-estrodiol (E