EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
Allosteric modulation of nicotinic and GABA
PMID 28842344
Abstract
Autism spectrum disorder (ASD) is associated with two core symptoms (social communication deficits and stereotyped repetitive behaviors) in addition to a number of comorbidities. There are no FDA-approved drugs for the core symptoms and the changes that underlie these behaviors are not fully understood. One hypothesis is an imbalance of the excitation (E)/inhibition (I) ratio with excessive E and diminished I occurring in specific neuronal circuits. Data suggests that both gamma-aminobutyric acid
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
631949
Methyl 4-hydroxy-3-iodobenzoate, 97%
C8H7IO3