EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Switching of the substrate specificity of protein tyrosine phosphatase N12 by cyclin-dependent kinase 2 phosphorylation orchestrating 2 oncogenic pathways.
PMID 28842430
Abstract
The protein tyrosine phosphatase nonreceptor type 12 (PTPN12) is a multifunctional protein and has elicited much research attention because its decreased protein level has been associated with poor prognosis of several types of cancers. Recently, we have solved the crystal structure of the phosphatase domain of PTPN12, which disclosed a specific PTPN12-insert-loop harboring a cyclin-dependent kinase 2 (CDK2) phosphorylation site. However, the functional significance of this phosphorylation is undefined. In the present study, we found that S19 site phosphorylation of PTPN12 by CDK2 discharged its antitumor activity by down-regulation of its inhibitory role in cell migration, but not affecting its other regulatory functions. Phosphorylation of PTPN12 at the S19 site changed its substrate interface, and by doing so, selectively decreased its activity toward the human epidermal growth factor receptor 2 (HER2)- pY