EMAIL THIS PAGE TO A FRIEND
Journal of toxicology. Clinical toxicology
Acute hallucinogenic reaction to carbinoxamine maleate.
PMID 2884327
Abstract
A reaction to Rondec is reported. The patient presented with agitation and hallucinations and responded promptly to a trial of physostigmine. The clinical picture and prompt resolution of symptoms with physostigmine strongly suggest that the reaction was to the antihistamine component of the medication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8278
Carbinoxamine maleate salt, analytical standard
C16H19ClN2O · C4H4O4