EMAIL THIS PAGE TO A FRIEND
Environmental research
Imbalanced immune responses involving inflammatory molecules and immune-related pathways in the lung of acute and subchronic arsenic-exposed mice.
PMID 28843991
Abstract
Inorganic arsenic has been claimed to increase the risk of pulmonary diseases through ingestion, as opposed to inhalation, which makes it a unique and intriguing environmental toxicant. However, the immunotoxic effects of lung, one of the targets of arsenic exposure, have not been extensively investigated in vivo. In the present study, we first confirmed that 2.5, 5 and 10mg/kg NaAsO