EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Nano-sized titanium dioxide toxicity in rat prostate and testis: Possible ameliorative effect of morin.
PMID 28844848
Abstract
This study investigated the effect of short-term oral exposure to nano-sized titanium dioxide (nTiO