EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Gadolinium Doping in Zirconia-Toughened Alumina Systems and Their Structural, Mechanical, and Aging Behavior Repercussions.
PMID 28846397
Abstract
A series of Gd