EMAIL THIS PAGE TO A FRIEND
Health physics
Performance Testing Of Selected Types of Electronic Personal Dosimeters in X- and Gamma Radiation Fields.
PMID 28846586
Abstract
Electronic personal dosimeters (EPDs) are increasingly being used alongside conventional thermoluminescent dosimeters to measure the dose of legal record in terms of personal dose equivalent. Therefore, it is of great importance to execute performance tests of these dosimeters in photon fields of various energies and at various angles of incidence. This testing is done in order to simulate the behavior of these dosimeters in realistic multidirectional polyenergetic ionizing radiation fields. Tests of accuracy, linearity, energy response, and angular response have been performed on 10 EPDs from multiple manufacturers. Various radiation qualities have been used in the energy range from 33 keV to 1.33 MeV and for angles of incidence 0° to 80°. This research proves that many of the EPDs tested performed according to the manufacturer's specifications and the requirements of the international standards regarding personal dosimetry.