EMAIL THIS PAGE TO A FRIEND
Alcohol (Fayetteville, N.Y.)
Is catalase involved in the effects of systemic and pVTA administration of 4-methylpyrazole on ethanol self-administration?
PMID 28847383
Abstract
The oxidative metabolism of ethanol into acetaldehyde involves several enzymes, including alcohol dehydrogenase (ADH) and catalase-hydrogen peroxide (H