EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
The effect of brassinolide, a plant steroid hormone, on drug resistant small-cell lung carcinoma cells.
PMID 28847728
Abstract
Small-cell lung carcinoma (SCLC) has a dismal prognosis in part because of multidrug resistance (MDR). Epibrassinolide (EB) is a steroid hormone in plants, with many physiological effects. It acts via a membrane receptor and GSK3 pathway, resulting in stabilization of a transcription factor. The parallels to the Wnt signaling pathway, which is activated in SCLC and results in increased β-catenin, prompted investigations of the effects of EB on drug-resistant (VPA17) and drug-sensitive (H69) SCLC cells. EB was cytotoxic to both cell lines (IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1641
Epibrassinolide, ≥85%
C28H48O6