EMAIL THIS PAGE TO A FRIEND
The Journal of experimental medicine
Dendritic cell and antigen dispersal landscapes regulate T cell immunity.
PMID 28847868
Abstract
Dendritic cell (DC) subsets with biased capacity for CD4