EMAIL THIS PAGE TO A FRIEND
Cancer immunology research
Neoadjuvant Interferons: Critical for Effective PD-1-Based Immunotherapy in TNBC.
PMID 28848054
Abstract
The lack of targeted therapies available for triple-negative breast cancer (TNBC) patients who fail to respond to first-line chemotherapy has sparked interest in immunotherapeutic approaches. However, trials utilizing checkpoint inhibitors targeting the PD-1/PD-L1 axis in TNBC have had underwhelming responses. Here, we investigated the interplay between type I IFN signaling and the PD-1/PD-L1 axis and tested the impact of combining IFN inducers, as immune activators, with anti-PD-1, to induce an antimetastatic immune response. Using models of TNBC, we demonstrated an interplay between type I IFN signaling and tumor cell PD-L1 expression that affected therapeutic response. The data revealed that the type I IFN-inducer poly(I:C) was an effective immune activator and antimetastatic agent, functioning better than anti-PD-1, which was ineffective as a single agent. Poly(I:C) treatment induced PD-L1 expression on TNBC cells, and combined poly(I:C) and anti-PD-1 treatment prolonged metastasis-free survival in a neoadjuvant setting via the induction of a tumor-specific T-cell response. Use of this combination in a late treatment setting did not impact metastasis-free survival, indicating that timing was critical for immunotherapeutic benefit. Together, these data demonstrated anti-PD-1 as an ineffective single agent in preclinical models of TNBC. However, type I IFN inducers were effective immune activators, and neoadjuvant trials combining them with anti-PD-1 to induce a sustained antitumor immune response are warranted.