EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Synthesis of p-nitrophenyl 5-acetamido-3,5-dideoxy-alpha-D-glycero-D-galacto-2-nonulopyranosid onic acid, a chromogenic substrate for sialidases.
PMID 2885090
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1516
2-O-(p-Nitrophenyl)-α-D-N-acetylneuraminic acid, ≥95%
C17H22N2O11