EMAIL THIS PAGE TO A FRIEND
Iranian journal of basic medical sciences
Crocin has anti-inflammatory and protective effects in ischemia-reperfusion induced renal injuries.
PMID 28852439
Abstract
Crocus sativus (saffron) has been widely used in traditional medicine. It has also been found to possess many beneficial properties in modern medicine. The most important ingredients of saffron are crocin, crocetin, safranal, and picrocrocin. This study evaluated the protective effects of crocin against the inflammation, oxidative stress, and functional disturbances of the kidney induced by renal ischemia/reperfusion (I/R). Different doses of crocin (0, 100, 200, and 400 mg/kg) were administered intraperitoneally 30 min before I/R. The rats of the sham group were also injected with normal saline before the sham surgery. For induction of I/R, both renal artery and vein clamped for 30 min, bilaterally. The I/R-induced renal injuries were assessed by measuring leukocyte infiltration, intercellular adhesion molecule-1 (ICAM-1) and tumor necrosis factor-alpha (TNF-α) mRNA expression levels, malondialdehyde (MDA) and ferric reducing/antioxidant power (FRAP) levels in the kidney tissue, and plasma creatinine and urea-nitrogen concentrations. Except for the tissue level of FRAP which decreased, all other measured parameters increased following I/R induction. Pretreatment with all doses of crocin significantly reduced the severity of these disturbances ( The present study clearly demonstrated the anti-inflammatory, antioxidant, and protective effects of crocin, a main constituent of saffron, against renal damages resulted from I/R in rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17304
Crocin, for microscopy
C44H64O24