EMAIL THIS PAGE TO A FRIEND
Aging
Aging effects on intestinal homeostasis associated with expansion and dysfunction of intestinal epithelial stem cells.
PMID 28854151
Abstract
Intestinal epithelial stem cells (IESCs) are critical to maintain intestinal epithelial function and homeostasis. We tested the hypothesis that aging promotes IESC dysfunction using old (18-22 months) and young (2-4 month) Sox9-EGFP IESC reporter mice. Different levels of Sox9-EGFP permit analyses of active IESC (Sox9-EGFP