EMAIL THIS PAGE TO A FRIEND
The Journal of experimental biology
Pharmacological effects of cannabinoids on learning and memory in
PMID 28855319
Abstract
Cannabinoids are hypothesized to play an important role in modulating learning and memory formation. Here, we identified mRNAs expressed in
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6226
AM251, >98% (HPLC), solid
C22H21N4OCl2I