EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
NQO-Induced DNA-Less Cell Formation Is Associated with Chromatin Protein Degradation and Dependent on A
PMID 28855893
Abstract
To investigate DNA damage response in the model crenarchaeon