EMAIL THIS PAGE TO A FRIEND
Biomedical optics express
Visualization of laser tattoo removal treatment effects in a mouse model by two-photon microscopy.
PMID 28856046
Abstract
Laser tattoo removal is an effective method of eliminating tattoo particles in the skin. However, laser treatment cannot always remove the unwanted tattoo completely, and there are risks of either temporary or permanent side effects. Studies using preclinical animal models could provide detailed information on the effects of laser treatment in the skin, and might help to minimize side effects in clinical practices. In this study, two-photon microscopy (TPM) was used to visualize the laser treatment effects on tattoo particles in both phantom specimens and
Related Materials