EMAIL THIS PAGE TO A FRIEND
Biomedical optics express
Toward whole-body quantitative photoacoustic tomography of small-animals with multi-angle light-sheet illuminations.
PMID 28856049
Abstract
Several attempts to achieve the quantitative photoacoustic tomography (q-PAT) have been investigated using point sources or a single-angle wide-field illumination. However, these schemes normally suffer from low signal-to-noise ratio (SNR) or poor quantification in imaging applications on large-size domains, due to the limitation of ANSI-safety incidence and incompleteness in the data acquisition. We herein present a q-PAT implementation that uses multi-angle light-sheet illuminations and calibrated recovering-and-averaging iterations. The scheme can obtain more complete information on the intrinsic absorption from the multi-angle illumination mode, and collect SNR-boosted photoacoustic signals in the selected planes from the wide-field light-sheet excitation. Therefore, the sliced absorption maps over whole body of small-animals can be recovered in a measurement-flexible, noise-robust and computation-economic way. The proposed approach is validated by phantom,
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9414
Agarose, low gelling temperature, BioReagent, for molecular biology