EMAIL THIS PAGE TO A FRIEND
Experimental & molecular medicine
Serum amyloid A inhibits dendritic cell differentiation by suppressing GM-CSF receptor expression and signaling.
PMID 28857084
Abstract
In this study, we report that an acute phase reactant, serum amyloid A (SAA), strongly inhibits dendritic cell differentiation induced by GM-CSF plus IL-4. SAA markedly decreased the expression of MHCII and CD11c. Moreover, SAA decreased cell surface GM-CSF receptor expression. SAA also decreased the expression of PU.1 and C/EBPα, which play roles in the expression of GM-CSF receptor. This inhibitory response by SAA is partly mediated by the well-known SAA receptors, Toll-like receptor 2 and formyl peptide receptor 2. Taken together, we suggest a novel insight into the inhibitory role of SAA in dendritic cell differentiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SRP6191 SAA1 monkey, recombinant, expressed in E. coli, ≥97% (SDS-PAGE)