EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Fingerprint profiles of flavonoid compounds from different Psidium guajava leaves and their antioxidant activities.
PMID 28857481
Abstract
Flavonoids are the main active components in Psidium guajava leaves and have many multi-physiological functions. In this study, the flavonoid compositions were identified in the Psidium guajava leaves samples using a high-performance liquid chromatography with time-of-flight electrospray ionization mass spectrometry method. A high-performance liquid chromatography fingerprint method, combined with chemometrics, was used to perform a quality assessment of the Psidium guajava leaves samples. The eight identified flavonoid compounds including rutin, isoquercitrin, quercetin-3-O-β-d-xylopyranoside, quercetin-3-O-α-l-arabinopyranoside, avicularin, quercitrin, quercetin, and kaempferol were used as the chemical markers. The antioxidant activity of 15 batches of samples was examined using three different methods, and the results revealed the Psidium guajava leaves samples that had higher contents of the flavonoid compounds, glycoside and aglycone, possessed the highest antioxidant capacities. Consequently, a combination of chromatographic fingerprints and chemometric analyses was used for a quality assessment of Psidium guajava leaf tea and its derived products, which can lay the foundation for the development of plant tea resources or other herbs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00740580
Quercitrin, primary reference standard
C21H20O11