EMAIL THIS PAGE TO A FRIEND
Enzyme and microbial technology
Coupling xylitol dehydrogenase with NADH oxidase improves l-xylulose production in Escherichia coli culture.
PMID 28859803
Abstract
Escherichia coli expressing NAD-dependent xylitol-4-dehydrogenase (XDH) from Pantoea ananatis and growing on glucose or glycerol converts xylitol to the rare sugar l-xylulose. Although blocking potential l-xylulose consumption (l-xylulosekinase, lyxK) or co-expression of the glycerol facilitator (glpF) did not significantly affect l-xylulose formation, co-expressing XDH with water-forming NADH oxidase (NOX) from Streptococcus pneumoniae increased l-xylulose formation in shake flasks when glycerol was the carbon source. Controlled batch processes at the 1L scale demonstrated that the final equilibrium l-xylulose/xylitol ratio was correlated to the intracellular NAD