EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
High serum adipocyte fatty acid binding protein level as a potential biomarker of aortic arterial stiffness in hypertensive patients with metabolic syndrome.
PMID 28860092
Abstract
The adipocyte-fatty-acid-binding protein (A-FABP) has been implicated in arterial stiffness, metabolic syndrome (MetS), and cardiovascular disease. We aimed to determine the relationship among serum A-FABP concentration, cardiometabolic risk factors, and central arterial stiffness in a hypertensive population. Fasting blood samples and baseline characteristics were obtained from 110 hypertensive patients. Serum A-FABP concentrations were determined by enzyme immunoassay kit. High arterial stiffness was defined as carotid-femoral pulse wave velocity values >10m/s via the SphygmoCor system. Patients with MetS and high arterial stiffness accounted for 67.3% and 42.7% of the study population, respectively. Serum A-FABP was positively associated with MetS and high arterial stiffness (P=0.006 and P<0.001, respectively). Multivariable stepwise linear regression analysis of the significant variables of arterial stiffness revealed that logarithmically transformed A-FABP (log-A-FABP, β=0.278, P=0.002) was positively correlated arterial stiffness in hypertensive patients. Subgroup analysis revealed that log-A-FABP (β=0.327, P=0.003), age (β=0229, P=0.032), and triglyceride (β=0.307, P=0.004) were significantly positively correlated with arterial stiffness in hypertensive patients with MetS. Elevated A-FABP concentration could be a predictor for MetS and arterial stiffness in hypertensive patients.