EMAIL THIS PAGE TO A FRIEND
The Journal of experimental biology
Strategies of ionoregulation in the freshwater nymph of the mayfly
PMID 28860119
Abstract
This study investigated ionoregulatory strategies used by freshwater (FW) nymphs of the mayfly