EMAIL THIS PAGE TO A FRIEND
Plant physiology
Seasonal Regulation of Petal Number.
PMID 28860156
Abstract
Four petals characterize the flowers of most species in the Brassicaceae family, and this phenotype is generally robust to genetic and environmental variation. A variable petal number distinguishes the flowers of