EMAIL THIS PAGE TO A FRIEND
Drug design, development and therapy
Paeoniflorin suppresses pancreatic cancer cell growth by upregulating HTRA3 expression.
PMID 28860718
Abstract
Paeoniflorin (PF) is an active monoterpene glycoside extracted from