EMAIL THIS PAGE TO A FRIEND
Ecology and evolution
Populations genetically rifting within a complex geological system: The case of strong structure and low genetic diversity in the migratory freshwater catfish,
PMID 28861223
Abstract
The complex geological history of East Africa has been a driving factor in the rapid evolution of teleost biodiversity. While there is some understanding of how macroevolutionary drivers have shaped teleost speciation in East Africa, there is a paucity of research into how the same biogeographical factors have affected microevolutionary processes within lakes and rivers. To address this deficiency, population genetic diversity, demography, and structure were investigated in a widely distributed and migratory (potamodromous) African teleost species, Ssemutundu (