EMAIL THIS PAGE TO A FRIEND
Virologica Sinica
Recombinant rabies virus expressing IL-15 enhances immunogenicity through promoting the activation of dendritic cells in mice.
PMID 28861771
Abstract
Rabies remains a public health threat that kills approximately 59,000 people worldwide each year, most of which are from the developing countries of Africa and Asia where dog rabies are endemic. Therefore, developing an affordable and efficacious vaccine is crucial for rabies control in these countries. Interleukin (IL)-15, an immunoregulatory cytokine, is a pluripotent molecule with therapeutic potential, which targets many cell types and links the innate and adaptive immune system. In this study, IL-15 gene was cloned and inserted into the genome of a recombinant rabies virus (RABV) strain LBNSE (designated as LBNSE-IL15), and the effect of over-expression of IL-15 on the immunogenicity of RABV was investigated. It was found that mice vaccinated with LBNSE-IL15 could induce significantly higher level of virus-neutralizing antibody (VNA) than those immunized with LBNSE, resulting in the higher protection after challenge. Further investigation was performed to find out the possible role of IL-15 plays in the process of antibody induction, and it was found that LBNSE-IL15 could enhance the maturation of dendritic cells (DCs) in immunized mice. Furthermore, the mice immunized with LBNSE-IL15 could promote the T