EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
From Vegetable Waste to New Agents for Potential Health Applications: Antioxidant Properties and Effects of Extracts, Fractions and Pinocembrin from Glycyrrhiza glabra L. Aerial Parts on Viability of Five Human Cancer Cell Lines.
PMID 28862446
Abstract
Glycyrrhiza glabra cultivation and harvesting produces substantial quantities of aerial parts as waste. With the aim to prospect an innovative valorization of these byproducts, the aerial parts were harvested in May and October and analyzed for their chemical profile, antioxidant properties, and effects on viability of five cancer cell lines. Pinocembrin was the main constituent. A significant protection of lipid peroxidation was observed with the May total extract (IC