EMAIL THIS PAGE TO A FRIEND
Waste management (New York, N.Y.)
Crude ethanolic extract from spent coffee grounds: Volatile and functional properties.
PMID 28865904
Abstract
Espresso capsule consumption and spent coffee ground (SCG) generation have increased, and the present study was undertaken to evaluate the volatile profile (VP), the antioxidant activity (AA) and the sun protection factor (SPF) of the Crude ethanolic extract obtained from the SCG in capsules. The extract yield was superior to the ether yield because a higher unsaponifiable matter (U.M.) amount was recovered by ethanol. The obtained VP (70 compounds) was typical of roasted coffee oil. Furthermore, chemometric analysis using principal components (PCA) discriminated the extracts and grouped the replicates for each sample, which showed the repeatability of the extraction process. The AA ranged from 18.4 to 23.6 (mg extract mg DPPH
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W391915
2-Ethyl-6-methylpyrazine, mixture of isomers, ≥95%
C7H10N2